missing translation for 'onlineSavingsMsg'
Learn More

Naphthol AS-BI phosphate, 93%, Thermo Scientific Chemicals

Catalog Number p-3758541
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1919-91-1
C18H15BrNO6P
452.2
MFCD00004058
HUXIAXQSTATULQ-UHFFFAOYSA-N
naphthol as-bi phosphate, naphthol as bi phosphate, 7-bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, 2-naphthalenecarboxamide, 7-bromo-n-2-methoxyphenyl-3-phosphonooxy, naphthol as-bi phosphoric acid, naphthol as-bi phosphoric acid solution, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl oxyphosphonic acid, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl dihydrogen phosphate, 2-naphthalenecarboxamide,7-bromo-n-2-methoxyphenyl-3-phosphonooxy-, sodium salt 1:2, naphthol as-bl phosphate
74715
[6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl] dihydrogen phosphate
COC1=CC=CC=C1NC(=O)C2=C(C=C3C=CC(=CC3=C2)Br)OP(=O)(O)O
This item is not returnable. View return policy
1919-91-1
C18H15BrNO6P
452.2
MFCD00004058
HUXIAXQSTATULQ-UHFFFAOYSA-N
naphthol as-bi phosphate, naphthol as bi phosphate, 7-bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, 2-naphthalenecarboxamide, 7-bromo-n-2-methoxyphenyl-3-phosphonooxy, naphthol as-bi phosphoric acid, naphthol as-bi phosphoric acid solution, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl oxyphosphonic acid, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl dihydrogen phosphate, 2-naphthalenecarboxamide,7-bromo-n-2-methoxyphenyl-3-phosphonooxy-, sodium salt 1:2, naphthol as-bl phosphate
74715
[6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl] dihydrogen phosphate
COC1=CC=CC=C1NC(=O)C2=C(C=C3C=CC(=CC3=C2)Br)OP(=O)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 91.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 91% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: partly soluble
Formula Weight 452.2
Percent Purity 93%
Physical Form Crystalline Powder
Chemical Name or Material Naphthol AS-BI phosphate
Show More Show Less
compliance-icons
Product Identifier
  • Naphthol AS-BI phosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'