missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7055951
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
256652-04-7
C16H19BO2
254.136
MFCD05663868
SPPZBAGKKBHZRW-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane, naphthalene-2-boronic acid pinacol ester, naphthalene-2-boronic acid, pinacol ester, 2-naphthaleneboronic acid, pinacol ester, 2-naphth-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthalenyl, 2-naphthaleneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-naphthalenyl-1,3,2-dioxaborolane, amtb668
10753409
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3C=C2
This item is not returnable. View return policy
256652-04-7
C16H19BO2
254.136
MFCD05663868
SPPZBAGKKBHZRW-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane, naphthalene-2-boronic acid pinacol ester, naphthalene-2-boronic acid, pinacol ester, 2-naphthaleneboronic acid, pinacol ester, 2-naphth-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthalenyl, 2-naphthaleneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-naphthalenyl-1,3,2-dioxaborolane, amtb668
10753409
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3C=C2

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 254.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Naphthalene-2-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'