missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1,4-diboronic Acid (~90%), TRC

Product Code. 30469973
Change view
Click to view available options
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30469973 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469973 Supplier Toronto Research Chemicals Supplier No. TRCN37767810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Naphthalene-1,4-diboronic Acid
CAS 22871-75-6
Molekylær formel C10H10B2O4
Mængde 10 g
Synonym B,B'-(1,4-Naphthalenediyl)bis-boronic Acid, 1,4-Naphthalenediboronic Acid, 1,4-Naphthalenediylbis-boronic Acid, [4-(Dihydroxyboranyl)naphthalen-1-yl]boronic Acid
InChI formel InChI=1S/C10H10B2O4/c13-11(14)9-5-6-10(12(15)16)8-4-2-1-3-7(8)9/h1-6,13-16H
SMIL OB(O)c1ccc(B(O)O)c2ccccc12
IUPAC navn (4-borononaphthalen-1-yl)boronic acid
Molekylvægt (g/mol) 215.81
Formel vægt 216.0765
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.