missing translation for 'onlineSavingsMsg'
Få mere at vide

Naphthalene-1,3-diol, 98%, BLD Pharm™

Artikelnummer. 31089801
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
250 mg
1 g
5 g
10 g
25 g
100 g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31089801 100 mg
31076173 100 g
31083035 10 g
31085852 250 mg
31088990 5 g
31078928 1 g
31081293 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31089801 Leverandør BLD Pharm Leverandørnr. BD12743100mg
 Begrænset vare
Forventet forsendelse: 25-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale Naphthalene-1,3-diol
CAS 132-86-5
Molekylær formel C10H8O2
MDL nummer MFCD00003965
Mængde 100 mg
SMIL OC1=C2C=CC=CC2=CC(O)=C1
Molekylvægt (g/mol) 160.17
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.