Få mere at vide
Tocris Bioscience™ Napabucasin
STAT3 inhibitor; also blocks cancer stem cell self-renewal
Brand: Tocris Bioscience™ 5522/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Napabucasin is a STAT3 inhibitor. Blocks spherogenesis of cancer stem cells (CSC), but not hematopoietic stem cells. Inhibits PaCa-2 xenograft tumor growth in mice, even after cessation of treatment, and decreases population of CSCs in PaCa-2 tumors from treated mice. Also blocks spleen and liver metastases of colon cancer cells in a mouse model.Tekniske data
| 2-Acetylnaphtho[2,3-b]furan-4,9-dione | |
| 83280-65-3 | |
| 10 mg |