missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Napabucasin

Description
Napabucasin is a STAT3 inhibitor. Blocks spherogenesis of cancer stem cells (CSC), but not hematopoietic stem cells. Inhibits PaCa-2 xenograft tumor growth in mice, even after cessation of treatment, and decreases population of CSCs in PaCa-2 tumors from treated mice. Also blocks spleen and liver metastases of colon cancer cells in a mouse model.
Specifications
Specifications
| Kemisk navn eller materiale | 2-Acetylnaphtho[2,3-b]furan-4,9-dione |
| CAS | 83280-65-3 |
| Mængde | 50 mg |
| Mål | STAT Inhibitors |
| Molekylær formel | C14H8O4 |
| Formel vægt | 240.21 |
| Renhed | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?