Få mere at vide
Greiner Bio-One NanoShuttle-PL Refill
Artikelnummer. 15800715
Skift visning
Mængde:
3 x 600 μl-hætteglas
6 x 600 μl-hætteglas
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15800715 3 x 600 μl-hætteglas
15817656 6 x 600 μl-hætteglas
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 15800715 Leverandør Greiner Bio-One Leverandørnr. 657843
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Suitable for use with magnetic 3D cell culturing.
  • Gold, iron oxide, and poly-L-lysine
  • Magnetizes cells by electrostatically attaching to cell membranes during an overnight static incubation
  • Magnetized cells will appear peppered with dark nanoparticles after incubation
  • Will stay attached to the cell membrane for up to 8 days, at which point it’s released into the 3D culture.
  • Biocompatible, having no effect on metabolism, proliferation, and inflammatory stress, and even encouraging proliferation in 3D
  • Does not interfere with experimental techniques, such as fluorescence, or western blotting.
TRUSTED_SUSTAINABILITY
Tekniske data
Med tilsætningsstoffer Gold, Iron Oxide, Poly-L-lysine
Til brug med (applikation) Magnetic 3D Cell Culturing
Mængde 3 x 600 μl-hætteglas