Få mere at vide
Greiner Bio-One NanoShuttle-PL Refill
Beskrivelse
- Gold, iron oxide, and poly-L-lysine
- Magnetizes cells by electrostatically attaching to cell membranes during an overnight static incubation
- Magnetized cells will appear peppered with dark nanoparticles after incubation
- Will stay attached to the cell membrane for up to 8 days, at which point it’s released into the 3D culture.
- Biocompatible, having no effect on metabolism, proliferation, and inflammatory stress, and even encouraging proliferation in 3D
- Does not interfere with experimental techniques, such as fluorescence, or western blotting.
Tekniske data
Tekniske data
| Med tilsætningsstoffer | Gold, Iron Oxide, Poly-L-lysine |
| Til brug med (applikation) | Magnetic 3D Cell Culturing |
| Mængde | 3 x 600 μl-hætteglas |