Få mere at vide
Thermo Scientific™ NanoDrop Lite Plus Spectrophotometers
Quickly and easily assess sample concentration and purity ratios with the entry-level Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer. The instrument delivers accurate measurements in an easy-to-use, affordable package. Ideal for teaching and busy research labs.
Brand: Thermo Scientific™ NDLPLUSGL
Se flere versioner af dette produkt
Beskrivelse
- Standard measurements
- Measures purified DNA, RNA, and protein concentration up to 30 Abs
- Up to 1,500 ng/µL dsDNA
- Up to 1,200 ng/µL RNA
- Up to 45 mg/mL BS
- Calculates critical A260/A280 and A260/A230 purity ratios
- Measures purified DNA, RNA, and protein concentration up to 30 Abs
- Fast, accurate measurements from only 1–2 µL—saves precious samples, no waste
- Sample-retention technology uses surface tension to hold the sample in place—no cuvettes or expensive consumables are needed
- Intuitive interface
- Built-in touchscreen operation
- No computer required
- Data storage and transfer
- Last 1,000 measurements saved locally; export data to a USB device
- Optional printer available for freezer-compatible labels (not included)
- Compact and portable—fits on any lab bench, ready for your next experiment
- Affordably priced—stretch tight budgets
- Academic teaching—educate students on molecular biology principles and experimental techniques on an industry-leading instrument
- Academic research labs—facilitate your research and publication of your novel research findings
- Biotech start-ups—accelerate the develop of new products
Tekniske data
| 3% at 0.97 A, 302 nm, 23 ± 2°C Abs/mm at 25 °C | |
| UL/CSA and CE | |
| 1,500 ng/μL (1,200 ng/μL) dsDNA (RNA)45 mg/mL (21 mg/mL) BSA (lgG) | |
| 22 cm | |
| 2.0 ng/μL (1.6 ng/μL) dsDNA (RNA)0.06 mg/mL (0.03 mg/mL) BSA (lgG) | |
| LCD Touchscreen | |
| Microvolume | |
| 100/240 V | |
| 27 x 22 x 22 cm (0.6 x 8.7 x 8.7 in); W x D X H | |
| 22 cm | |
| ≤ 5 seconds | |
| 1.0 mm and 0.2 mm (auto adjust) | |
| No | |
| 1 to 2 μL | |
| Standalone control with data export via USB Flash Drive | |
| ± 1 nm | |
| 6.0 lbs (without printer) | |
| 27 cm |
| 0.04 to 30 Abs (10 mm equivalent) | |
| Local Control Only | |
| USB 2.0 Port | |
| NanoDrop Lite Plus Spectrophotometer (no printer) | |
| 2048-element CMOS Linear Image Sensor | |
| Xenon Flash | |
| Microvolume UV-Vis Spectrophotometer | |
| 230 nm, 260 nm, 280 nm | |
| Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification | |
| Stainless Steel (303), Quartz Fiber | |
| 1 | |
| 18 W (standby < 3 Watts) | |
| Standalone control with data export via USB Flash Drive | |
| ≤ 1.8 nm (FWHM at Hg 254 nm) | |
| One year | |
| Typical: 0.002 A (1.0 mm path) or 1%CV, whichever is greater. (SD of 20 individual measurements at 0.74 Abs) | |
| 2.7 kg (without printer) |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.