Få mere at vide
Nandrolone Phenylpropionate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Nandrolone Phenpropionate |
| CAS | 62-90-8 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C27 H34 O3 |
| Mængde | 10 mg |
| Synonym | Nandrolone Phenpropionate, Estr-4-en-3-one','17-(1-oxo-3-phenylpropoxy)-','(17β)-, Estr-4-en-3-one','17β-hydroxy-','hydrocinnamate (7CI,8CI), Estr-4-en-3-one','17β-hydroxy-','phenylpropionate (6CI), Hydrocinnamic acid','ester with 17β-hydroxyestr-4-en-3-one (8CI), 17β-(3-Phenylpropionyloxy)-estr-4-en-3-one, 17β-Hydroxy-19-norandrost-4-en-3-one 17-phenylpropionate, 17β-Hydroxy-4-estren-3-one17-phenylpropionate, 17β-Hydroxyestr-4-en-3-one phenylpropionate, 19-Nor-17β-hydroxy-3-ketoandrost-4-ene 17-phenylpropionate, 19-Norandrostenolone phenylpropionate, 19-Nortestosterone phenylpropionate, Activin (anabolic steroid), Durabol, Durabolin, Fenobolin, NPP, NSC 23162, NTPP, Nandrobolic, Nandrolin, Nandrolon phenylpropionate, Nandrolone phenpropionate, Nandrolone phenylpropionate, Nerobil, Nerobolil, Norandrolone phenylpropionate, Norandrostenolone phenylpropionate, Nortestosterone phenylpropionate, Phenobolin, Strabolene, Superanabolon |
| InChI formel | InChI=1S/C27H34O3/c1-27-16-15-22-21-11-9-20(28)17-19(21)8-10-23(22)24(27)12-13-25(27)30-26(29)14-7-18-5-3-2-4-6-18/h2-6,17,21-25H,7-16H2,1H3/t21-,22+,23+,24-,25-,27-/m0/s1 |
| SMIL | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@H]2OC(=O)CCc5ccccc5 |
| IUPAC navn | [(8R,9S,10R,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,9,10,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl] 3-phenylpropanoate |
| Molekylvægt (g/mol) | 406.56 |
| Vis mere |