Få mere at vide
Nandrolone Decanoate, LoGiCal
Artikelnummer. 30464197
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464197 10 mg
1 options
Artikelnummer. 30464197 Leverandør LoGiCal Leverandørnr. LGCFOR0982.00
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nandrolone Decanoate
CAS 360-70-3
Analyt- eller komponentnavne Nandrolone Decanoate
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C28 H44 O3
Certificeringer/compliance ISO 9001
Synonym Nandrolone Decanoate, Estr-4-en-3-one, 17-[(1-oxodecyl)oxy]-, (17beta)-, Estr-4-en-3-one, 17beta-hydroxy-, decanoate (6CI,7CI,8CI), Decanoic acid, ester with 17beta-hydroxyestr-4-en-3-one (8CI), 17beta-Hydroxy-19-norandrost-4-en-3-one 17-decanoate, 17beta-Hydroxyestr-4-en-3-one 17-decanoate, 17beta-Hydroxyestr-4-en-3-one decanoate, 17beta-Nandrolone laureate, 17beta-Nortestosterone laureate, 19-Nor-17beta-hydroxy-3-ketoandrost-4-ene 17-decanoate, 19-Norandrostenolone decanoate, 19-Nortestosterone 17beta-decanoate, 19-Nortestosterone decanoate, Anabolicum, Anabolin Forte, Deca-Durabol, Deca-Durabolin, Deca-Hybolin, Decanofort, Extraboline, Hybolin decanoate, Nandrobolic LA, Norandrostenolone decanoate, Nortestosterone decanoate, Retabolil
InChI formel InChI=1S/C28H44O3/c1-3-4-5-6-7-8-9-10-27(30)31-26-16-15-25-24-13-11-20-19-21(29)12-14-22(20)23(24)17-18-28(25,26)2/h19,22-26H,3-18H2,1-2H3/t22-,23+,24+,25-,26-,28-/m0/s1
Løsningstype Neat
SMIL CCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C
IUPAC navn [(8R,9S,10R,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,9,10,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl] decanoate
Industry Type Clinical Testing
Molekylvægt (g/mol) 428.647
Mængde 10 mg
Formel vægt 428.329 g/mol