Få mere at vide
Nandrolone-16,16,17-d3, CDN
Artikelnummer. 30318347
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318347 0.01 g
30310526 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318347 Leverandør CDN Leverandørnr. CDND57350.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nandrolone-d3
CAS 120813-22-1
Molekylær formel C18 D3 H23 O2
Mængde 0.01 g
Synonym Estr-4-en-3-one-16,16,17-d3, 17-hydroxy-, (17β)-, (17β)-17-Hydroxyestr-4-en-3-one-16,16,17-d3, Nandrolone-d3, d3-17β-Nandrolone, 19-Nortestosterone (16,16,17-D3), Nandrolone-16,16,17-D3
InChI formel InChI=1 S/C18H26O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h10,13-17,20 H,2-9H2,1H3/t13-,14+,15+,16-,17-,18-/m0/s1/i7D2,17 D
SMIL [2 H]C1([2 H])C[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@@H]4 [C@H]3 CC[C@]2(C)[C@@]1([2 H])O
IUPAC navn (8 R,9 S,10 R,13 S,14 S,17 S)-16,16,17-trideuterio-17-hydroxy-13-methyl-2,6,7,8,9,10,11,12,14,15-decahydro-1 H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 277.416
Formel vægt 277.212 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity