Få mere at vide
Namitecan, MedChemExpress
Artikelnummer. 16496188
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16496188 10 mg
16458787 5 mg
16421449 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16496188 Leverandør Medchem Express Leverandørnr. HY14821
MedChemExpress Namitecan is a potent topoisomerase I inhibitor, with antitumor property.
Namitecan is a potent topoisomerase I inhibitor, with antitumor property.
Tekniske data
Kemisk navn eller materiale Namitecan
Fysisk form Solid
CAS 372105-27-6
Farve Yellow
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C23H22N4O5
Til brug med (applikation) Neuroscience-Neuromodulation
Synonym ST-1968
Opløselighedsinformation DMSO : 250 mg/mL (575.45 mM; Need ultrasonic)
SMIL O=C1[C@](O)(CC)C2=C(CO1)C(N3CC4=C(/C=N/OCCN)C5=CC=CC=C5N=C4C3=C2)=O
Molekylvægt (g/mol) 434.44
Formel vægt 434.44
Procent renhed 98.11%