Få mere at vide
Nalpha-Phenyl-d5-acetyl-L-glutamine, CDN
Artikelnummer. 30312667
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312667 0.01 g
30316346 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312667 Leverandør CDN Leverandørnr. CDND69000.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phenyl-d5-acetyl L-Glutamine
CAS 1331909-01-3
Molekylær formel C13 D5 H11 N2 O4
Mængde 0.01 g
Synonym L-Glutamine, N2-[2-(phenyl-2,3,4,5,6-d5)acetyl]- (ACI), N2-[2-(Phenyl-2,3,4,5,6-d5)acetyl]-L-glutamine (ACI), (2 S)-5-Amino-5-oxo-2-[[2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]pentanoic acid, Nα-(Phenyl-d5-acetyl)-L-glutamine, Phenylacetyl-d5 L-Glutamine, (2 S)-5-Amino-5-oxo-2-[2-(phenyl-d5)acetamido]pentanoic acid, (2 S)-5-Amino-5-oxo-2-[2-(phenyl-d5)acetamido]pentanoic acid, (2 S)-5-Amino-5-oxo-2-[2-(phenyl-d5)acetamido]pentanoic acid
InChI formel InChI=1 S/C13H16N2O4/c14-11(16)7-6-10(13(18)19)15-12(17)8-9-4-2-1-3-5-9/h1-5,10 H,6-8H2,(H2,14,16)(H,15,17)(H,18,19)/t10-/m0/s1/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(CC(=O)N[C@@H](CCC(=O)N)C(=O)O)c([2 H])c1[2 H]
IUPAC navn (2 S)-5-amino-5-oxo-2-[[2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]pentanoic acid
Molekylvægt (g/mol) 269.308
Formel vægt 269.142 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity