missing translation for 'onlineSavingsMsg'
Learn More

Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals

Catalog Number p-7054519
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
15098-69-8
C28H53N3O6
527.75
MFCD00038892
HRLHJTYAMCGERD-VAMKTSTMNA-N
boc-lys boc-oh.dcha, boc-lys boc-oh dcha, dicyclohexylamine s-2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine dicyclohexylamine salt, boc-lys boc-oh dicyclohexammonium salt, n,n'-bis tert-butoxycarbonyl-l-lysine dicyclohexylamine 1:1, 2s-2,6-bis tert-butoxycarbonyl amino hexanoic acid; dicha, boc-lys boc-ohcha, boc-lys boc-oh?cha, boc-lys boc-ohdcha
12017189
(2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid;N-cyclohexylcyclohexanamine
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
This item is not returnable. View return policy
15098-69-8
C28H53N3O6
527.75
MFCD00038892
HRLHJTYAMCGERD-VAMKTSTMNA-N
boc-lys boc-oh.dcha, boc-lys boc-oh dcha, dicyclohexylamine s-2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine dicyclohexylamine salt, boc-lys boc-oh dicyclohexammonium salt, n,n'-bis tert-butoxycarbonyl-l-lysine dicyclohexylamine 1:1, 2s-2,6-bis tert-butoxycarbonyl amino hexanoic acid; dicha, boc-lys boc-ohcha, boc-lys boc-oh?cha, boc-lys boc-ohdcha
12017189
(2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid;N-cyclohexylcyclohexanamine
C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 139°C to 143°C
Color White
Beilstein 6262378
Formula Weight 527.74
Percent Purity 98%
Quantity 25 g
Physical Form Solid
Chemical Name or Material Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'