missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-D-lysine, 98%, Thermo Scientific Chemicals

Catalog Number 15459408
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
106719-44-2
C11H22N2O4
246.31
MFCD00076956
DQUHYEDEGRNAFO-SVGMAFHSNA-N
boc-d-lys-oh, boc-l-lys-oh, n2-boc-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-boc-d-lysine, n-, a-boc-d-lysine, boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine, n-alpha-t-butoxycarbonyl-d-lysine
7018770
(2R)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O
This item is not returnable. View return policy
106719-44-2
C11H22N2O4
246.31
MFCD00076956
DQUHYEDEGRNAFO-SVGMAFHSNA-N
boc-d-lys-oh, boc-l-lys-oh, n2-boc-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-boc-d-lysine, n-, a-boc-d-lysine, boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine, n-alpha-t-butoxycarbonyl-d-lysine
7018770
(2R)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 196°C to 200°C
Quantity 250 mg
Optical Rotation −22° (c=2 in Methanol)
Formula Weight 246.3
Percent Purity 98%
Chemical Name or Material Nalpha-Boc-D-lysine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'