missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-O-tert-butyl-L-serine, 98%

Brand:  Alfa Aesar™ H63904.14

Additional Details : CAS Number : 1676-75-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15400869

  • 1458.67 DKK / 25g


Estimated Shipment date
from Supplier 17-10-2018
Add to basket

Specifications

Chemical Identifiers

1676-75-1
295.335
TXDGEONUWGOCJG-LBPRGKRZSA-N
1715617
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C15H21NO5
MFCD00038275
Z-Ser(tBu)-OH; (S)-2-(Benzyloxycarbonylamino)-3-(tert-butoxy)propionic acid
(2S)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid

Specifications

Nalpha-Benzyloxycarbonyl-O-tert-butyl-L-serine
98%
MFCD00038275
Z-Ser(tBu)-OH; (S)-2-(Benzyloxycarbonylamino)-3-(tert-butoxy)propionic acid
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
295.335
295.33
25g
1676-75-1
C15H21NO5
2000284
TXDGEONUWGOCJG-LBPRGKRZSA-N
(2S)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid
1715617
Solid
Videos
SDS