Learn More
Alfa Aesar™ Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-serine, 98%
Brand: Alfa Aesar™ H63728.06
Additional Details : CAS Number : 65806-90-8 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
65806-90-8 | |
295.335 | |
TXDGEONUWGOCJG-GFCCVEGCSA-N | |
1715619 | |
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 |
C15H21NO5 | |
MFCD00077038 | |
z-d-ser tbu-oh, cbz-o-tert-butyl-d-serine, r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-cbz-o-tert-butyl-d-serine, 2r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, ambotzzaa1152, n-alpha-benzyloxycarbonyl-o-t-butyl-d-serine, n-alpha-carbobenzoxy-d-serine t-butyl ether, r-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-d-serine | |
(2R)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid |
Specifications
Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-serine | |
C15H21NO5 | |
z-d-ser tbu-oh, cbz-o-tert-butyl-d-serine, r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-cbz-o-tert-butyl-d-serine, 2r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, ambotzzaa1152, n-alpha-benzyloxycarbonyl-o-t-butyl-d-serine, n-alpha-carbobenzoxy-d-serine t-butyl ether, r-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-d-serine | |
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1 | |
295.335 | |
295.33 | |
98% | |
5g |
65806-90-8 | |
MFCD00077038 | |
TXDGEONUWGOCJG-GFCCVEGCSA-N | |
(2R)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid | |
1715619 | |
Powder | |
White |