Få mere at vide
Nalpha-Acetyl-DL-Glutamine-2,3,3,4,4-d5, CDN
Artikelnummer. 30320889
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320889 0.005 g
30315251 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320889 Leverandør CDN Leverandørnr. CDND81140.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N2-Acetyl-DL-glutamine-2,3,3,4,4-D5
CAS 14341-82-3
Molekylær formel C7 D5 H7 N2 O4
Mængde 0.005 g
Synonym Glutamine-2,3,3,4,4-d5, N2-acetyl-, DL- (8 CI), N2-Acetyl-DL-glutamine-2,3,3,4,4-D5, Nα-Acetyl-DL-glutamine-2,3,3,4,4-d5
InChI formel InChI=1 S/C7H12N2O4/c1-4(10)9-5(7(12)13)2-3-6(8)11/h5H,2-3H2,1H3,(H2,8,11)(H,9,10)(H,12,13)/i2D2,3D2,5 D
SMIL [2 H]C([2 H])(C(=O)N)C([2 H])([2 H])C([2 H])(NC(=O)C)C(=O)O
IUPAC navn 2-acetamido-5-amino-2,3,3,4,4-pentadeuterio-5-oxopentanoic acid
Molekylvægt (g/mol) 193.212
Formel vægt 193.111 g/mol
Procent renhed 98 atom % D, min 90% Chemical Purity