missing translation for 'onlineSavingsMsg'
Få mere at vide

Naloxone, TRC

Artikelnummer. 30007159
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007159 1 mg
30004154 10 mg
30010685 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30007159 Leverandør Toronto Research Chemicals Leverandørnr. TRCN2849951MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 465-65-6
Molekylær formel C19 H21 N O4
Molekylvægt (g/mol) 327.37
Synonym Morphinan-6-one, 4,5-epoxy-3,14-dihydroxy-17-(2-propen-1-yl)-, (5α)-, Morphinan-6-one, 17-allyl-4,5α-epoxy-3,14-dihydroxy- (8CI), Morphinan-6-one, 4,5-epoxy-3,14-dihydroxy-17-(2-propenyl)-, (5α)- (9CI), Normorphinone, N-allyl-7,8-dihydro-14-hydroxy- (7CI), (-)-Naloxone, 12-Allyl-7,7a,8,9-tetrahydro-3,7a-dihydroxy-4aH-8,9c-iminoethanophenanthro[4,5-bcd]furan-5(6H)-one, NSC 70413, Naloxone, l-Naloxone
IUPAC navn (4R,4aS,7aR,12bS)-4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one
SMIL Oc1ccc2C[C@H]3N(CC=C)CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O

Tekniske data

Kemisk navn eller materiale Naloxone
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C19H21NO4/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11/h2-4,14,17,21,23H,1,5-10H2/t14-,17+,18+,19-/m1/s1
Formel vægt 327.1471
Procent renhed >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.