Få mere at vide
Naloxegol Oxalate, TRC
Artikelnummer. 30132458
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132458 50 mg
30007435 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132458 Leverandør Toronto Research Chemicals Leverandørnr. TRCN28447550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1354744-91-4
Molekylær formel C34H53NO11 • C2H2O4
Molekylvægt (g/mol) 651.799003
Synonym AZ 13337019 Oxalate, Moventig, NKTR 118 Oxalate, (5α,6α)-4,5-Epoxy-6-(3,6,9,12,15,18,21-heptaoxadocos-1-yloxy)-17-(2-propen-1-yl)-morphinan-3,14-diol Ethanedioate
IUPAC navn (4R,4aS,7S,7aR,12bS)-7-[2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]-3-prop-2-enyl-1,2,4,5,6,7,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-4a,9-diol;oxalic acid
SMIL O[C@]12[C@]3(CCN(CC=C)[C@@H]2C4)[C@](OC5=C3C4=CC=C5O)([H])[C@@H](OCCOCCOCCOCCOCCOCCOCCOC)CC1.OC(C(O)=O)=O
Tekniske data
Kemisk navn eller materiale Naloxegol Oxalate
Mængde 50 mg
InChI formel InChI=1S/C34H53NO11.C2H2O4/c1-3-9-35-10-8-33-30-26-4-5-27(36)31(30)46-32(33)28(6-7-34(33,37)29(35)25-26)45-24-23-44-22-21-43-20-19-42-18-17-41-16-15-40-14-13-39-12-11-38-2;3-1(4)2(5)6/h3-5,28-29,32,36-37H,1,6-25H2,2H3;(H,3,4)(H,5,6)/t28-,29+,32-,33-,34+;/m0./s1
Formel vægt 741.3572