Learn More
Thermo Scientific™ Nalgene™ Stopcocks
Product Code. 10279070
Change view
Click to view available options
Plug Bore:
2 mm
4 mm
Unit Size:
Each
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Bore Unit Size
10279070 4 mm Each
15991651 2 mm Pack of 6
11369554 2 mm Pack of 6
15223079 4 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10279070 Supplier Thermo Scientific™ Supplier No. 64600004
  • ONLINE EXCLUSIVE PRICE
    390.00 DKK
    335.74 DKK / Pakke med 1
    Spare 54.26 DKK
    13% Slukket
View all promotions
Forventet forsendelse: 11-12-2026
Develop in-house fluid transfer systems for accurate filling or dispensing. These stopcocks provide reliable, accurate liquid flow control.
Permit reliable, accurate liquid flow control in liquid handling systems by using Thermo Scientific™ Nalgene™ Stopcocks. They are ideal for use in developing in-house fluid transfer systems for one-way fluid path for accurate filling or dispensing. The two-way stopcocks feature serrated tubulation on each end.
  • Serrated tubulation on each end accepts 1/2″ through 5/16″ tubing
  • Polypropylene with Teflon™ TFE plug
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Type Stopcock
Plug Bore 4 mm
Material Polypropylene
No. per Pack 1
No. per Case 6
For Use With (Equipment) In-House Fluid Transfer System
Plug Material Polypropylene
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.