Learn More
Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack
Package, ship and store a variety of reagents using these round, narrow-mouth LDPE bottles with closure.
Brand: Thermo Scientific™ 312003-0032
1439.97 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Package, ship and store a variety of reagents using Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure, constructed from natural low-density polyethylene.
Key Features- Durable and clear
- Uniform walls that resist splitting and puncturing
- Leakproof†when used with their Nalgene closures
- Ideal for trace metal analysis — LDPE is low in trace metal content
Specifications
32 oz., 1000 mL | |
32 oz. | |
Narrow-Mouth Boston Round Bottle | |
LDPE | |
50 | |
Natural | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
Narrow | |
216 mm | |
8.5 in. | |
Polypropylene | |
91 mm | |
No |
1 L | |
Natural | |
27 mm | |
50 | |
Non-sterile | |
38 mm | |
Yes | |
Round | |
Boston Round Bottle | |
Unlined | |
38-430 mm | |
Yes (Bulk Packed, Unassembled) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1415387 | Certificate of Compliance | 01/11/2024 | 312003-0032 |
1377061 | Certificate of Compliance | 28/01/2023 | 312003-0032 |
1373959 | Certificate of Compliance | 04/12/2022 | 312003-0032 |
1345675 | Certificate of Compliance | 19/02/2022 | 312003-0032 |
1309314 | Certificate of Compliance | 26/02/2021 | 312003-0032 |
†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.