Få mere at vide
Nahlsgen, TRC
Artikelnummer. 30475667
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475667 10 mg
30464899 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30475667 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49706310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 926281-37-0
Molekylær formel C13H18NO7P
Molekylvægt (g/mol) 331.26
Synonym 3-[[(3-Amino-3-carboxypropyl)methoxyphosphinyl]oxy]benzeneacetic Acid, GGsTop
IUPAC navn 2-amino-4-[[3-(carboxymethyl)phenoxy]-methoxyphosphoryl]butanoic acid
SMIL COP(=O)(CCC(N)C(=O)O)Oc1cccc(CC(=O)O)c1
Tekniske data
Kemisk navn eller materiale Nahlsgen
Mængde 10 mg
InChI formel InChI=1S/C13H18NO7P/c1-20-22(19,6-5-11(14)13(17)18)21-10-4-2-3-9(7-10)8-12(15)16/h2-4,7,11H,5-6,8,14H2,1H3,(H,15,16)(H,17,18)
Formel vægt 331.0821