Få mere at vide
Naftalofos, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Naftalofos |
| CAS | 1491-41-4 |
| Molekylær formel | C16 H16 N O6 P |
| Mængde | 250 mg |
| Synonym | 1H-Benz[de]isoquinoline-1,3(2H)-dione, 2-[(diethoxyphosphinyl)oxy]- (9CI), Naphthalimide, N-hydroxy-, diethyl phosphate (6CI,7CI,8CI), BAY 9002, Bayer 9002, ENT 25567, Maretin, N-Hydroxynaphthalimide diethyl phosphate, Naftalofos, Naphthalophos, O,O-Diethyl N-hydroxynaphthalimide phosphate, Rametin, S 940 |
| InChI formel | InChI=1S/C16H16NO6P/c1-3-21-24(20,22-4-2)23-17-15(18)12-9-5-7-11-8-6-10-13(14(11)12)16(17)19/h5-10H,3-4H2,1-2H3 |
| SMIL | CCOP(=O)(OCC)ON1C(=O)c2cccc3cccc(C1=O)c23 |
| IUPAC navn | (1,3-dioxobenzo[de]isoquinolin-2-yl) diethyl phosphate |
| Molekylvægt (g/mol) | 349.28 |
| Formel vægt | 349.0715 |