Få mere at vide
Nadolol EP Impurity F (as Hydrochloride), Mikromol™
Artikelnummer. 30052140
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30052140 25 mg
1 options
Artikelnummer. 30052140 Leverandør Mikromol Leverandørnr. MM0439.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nadolol EP Impurity F (as Hydrochloride)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-1-[(1,1-Dimethylethyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol Hydrochloride
CAS 15230-34-9
Analyt- eller komponentnavne (2RS)-1-[(1,1-Dimethylethyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C17 H23 N O2 . Cl H
Certificeringer/compliance ISO 17025
Synonym (2RS)-1-[(1,1-Dimethylethyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol Hydrochloride, Nadolol Imp. F (EP) as Hydrochloride, Nadolol Impurity F as Hydrochloride
InChI formel InChI=1S/C17H23NO2.ClH/c1-17(2,3)18-11-14(19)12-20-16-10-6-8-13-7-4-5-9-15(13)16;/h4-10,14,18-19H,11-12H2,1-3H3;1H
SMIL Cl.CC(C)(C)NCC(O)COc1cccc2ccccc12
IUPAC navn 1-(tert-butylamino)-3-naphthalen-1-yloxypropan-2-ol;hydrochloride
Molekylvægt (g/mol) 309.83
Mængde 25 mg
Formel vægt 309.1496 g/mol