Få mere at vide
Nadolol-d9, TRC
Artikelnummer. 30465113
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30465113 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465113 Leverandør Toronto Research Chemicals Leverandørnr. TRCN2010522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale iso-Nadolol (tert-Butyl-d9) (Mixture of Diastereomers)
CAS 1246820-10-9
Molekylær formel C17H18D9NO4
Mængde 2.5 mg
Synonym 5-[3-[(1,1-Dimethylethyl-d9)amino]-2-hydroxypropoxy]-1,2,3,4-tetrahydro-2,3-naphthalenediol, Anabet-d9, Corgard-d9, SQ 11725-d9, Solgol-d9
InChI formel InChI=1S/C17H27NO4/c1-17(2,3)18-9-13(19)10-22-14-5-4-11-7-15(20)16(21)8-12(11)6-14/h4-6,13,15-16,18-21H,7-10H2,1-3H3/i1D3,2D3,3D3
SMIL OC1C(O)CC(C=CC(OCC(O)CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=C2)=C2C1
IUPAC navn 6-[3-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-2-hydroxypropoxy]-1,2,3,4-tetrahydronaphthalene-2,3-diol
Molekylvægt (g/mol) 318.46
Formel vægt 318.2505