Få mere at vide
Nabumetone EP Impurity B, Mikromol™
Artikelnummer. 30457672
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30457672 4 x 25 mg
30052221 25 mg
17972696 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30457672 Leverandør Mikromol Leverandørnr. MM0153.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nabumetone EP Impurity B, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (5RS)-5-(6-Methoxynaphthalen-2-yl)-3-methylcyclohex-2-enone
CAS 343272-51-5
Analyt- eller komponentnavne (5RS)-5-(6-Methoxynaphthalen-2-yl)-3-methylcyclohex-2-enone
Forsendelsestilstand Room Temperature
Molekylær formel C18 H18 O2
Certificeringer/compliance ISO 17025
Synonym (5RS)-5-(6-Methoxynaphthalen-2-yl)-3-methylcyclohex-2-enone, Nabumetone Imp. B (EP), Nabumetone Impurity B
InChI formel InChI=1S/C18H18O2/c1-12-7-16(10-17(19)8-12)14-3-4-15-11-18(20-2)6-5-13(15)9-14/h3-6,8-9,11,16H,7,10H2,1-2H3
Løsningstype Neat
SMIL COc1ccc2cc(ccc2c1)C3CC(=CC(=O)C3)C
IUPAC navn 5-(6-methoxynaphthalen-2-yl)-3-methylcyclohex-2-en-1-one
Molekylvægt (g/mol) 266.33
Mængde 4 x 25 mg
Formel vægt 266.1307 g/mol