Få mere at vide
Nabumetone EP Impurity A, Mikromol™
Artikelnummer. 30049772
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049772 25 mg
30051423 100 mg
30457955 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30049772 Leverandør Mikromol Leverandørnr. MM0153.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nabumetone EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(6-Methoxynaphthalen-2-yl)-5-methylcyclohexanone
CAS 343272-52-6
Analyt- eller komponentnavne 3-(6-Methoxynaphthalen-2-yl)-5-methylcyclohexanone
Forsendelsestilstand Room Temperature
Molekylær formel C18 H20 O2
Certificeringer/compliance ISO 17025
Synonym 3-(6-Methoxynaphthalen-2-yl)-5-methylcyclohexanone, Nabumetone Imp. A (EP), Nabumetone Impurity A
InChI formel InChI=1S/C18H20O2/c1-12-7-16(10-17(19)8-12)14-3-4-15-11-18(20-2)6-5-13(15)9-14/h3-6,9,11-12,16H,7-8,10H2,1-2H3
SMIL COc1ccc2cc(ccc2c1)C3CC(C)CC(=O)C3
IUPAC navn 3-(6-methoxynaphthalen-2-yl)-5-methylcyclohexan-1-one
Molekylvægt (g/mol) 268.35
Mængde 25 mg
Formel vægt 268.1463 g/mol