Få mere at vide
N6-Succinyl Adenosine, TRC
Artikelnummer. 30009754
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009754 5 mg
30009253 1 mg
30008005 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009754 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6888255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4542-23-8
Molekylær formel C14 H17 N5 O8
Molekylvægt (g/mol) 383.31
Synonym N-(9-β-D-Ribofuranosyl-9H-purin-6-yl)-L-aspartic Acid, 6-(1,2-Dicarboxyethylamino)
IUPAC navn (2S)-2-[[9-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]amino]butanedioic acid
SMIL OC[C@H]1O[C@@H]([C@H](O)[C@@H]1O)n2cnc3c(N[C@@H](CC(=O)O)C(=O)O)ncnc23
Tekniske data
Kemisk navn eller materiale N6-Succinyl Adenosine
Mængde 5 mg
InChI formel InChI=1S/C14H17N5O8/c20-2-6-9(23)10(24)13(27-6)19-4-17-8-11(15-3-16-12(8)19)18-5(14(25)26)1-7(21)22/h3-6,9-10,13,20,23-24H,1-2H2,(H,21,22)(H,25,26)(H,15,16,18)/t5-,6+,9+,10+,13-/m0/s1
Formel vægt 383.1077