Få mere at vide
N6-Methyl-N6-threonylcarbamoyladenosine, TRC
Artikelnummer. 30134409
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134409 10 mg
30133379 5 mg
30010109 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134409 Leverandør Toronto Research Chemicals Leverandørnr. TRCM69625510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 39667-81-7
Molekylær formel C16 H22 N6 O8
Molekylvægt (g/mol) 426.38
Synonym (2S,3R)-3-Hydroxy-2-(3-(9-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-9H-purin-6-yl)-3-methylureido)butanoic Acid, N-[[Methyl(9-β-D-ribofuranosyl-9H-purin-6-yl)amino]carbonyl]-L-threonine, N-[(9-β-D-Ribofuranosylpurin-6-yl)-N-methylcarbamoyl]threonine, N-[N-Methyl-N-(9β-D-ribofuranosylpurin-6-yl)carbamoyl]threonine
IUPAC navn (2S,3R)-2-[[[9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl]purin-6-yl]-methyl-carbamoyl]amino]-3-hydroxy-butanoic acid
SMIL C[C@@H](O)[C@H](NC(=O)N(C)c1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)O
Tekniske data
Kemisk navn eller materiale N6-Methyl-N6-threonylcarbamoyladenosine
Mængde 10 mg
InChI formel InChI=1S/C16H22N6O8/c1-6(24)8(15(27)28)20-16(29)21(2)12-9-13(18-4-17-12)22(5-19-9)14-11(26)10(25)7(3-23)30-14/h4-8,10-11,14,23-26H,3H2,1-2H3,(H,20,29)(H,27,28)/t6-,7-,8+,10-,11-,14-/m1/s1
Formel vægt 426.1499