Få mere at vide
N6-Carbobenzoxy-L-lysine N-Carboxyanhydride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N6-Carbobenzoxy-L-lysine N-Carboxyanhydride |
| CAS | 1676-86-4 |
| Molekylær formel | C15 H18 N2 O5 |
| Mængde | 1 g |
| Synonym | Carbamic acid, N-[(4S)-4-(2,5-dioxo-4-oxazolidinyl)butyl]-, phenylmethyl ester, Carbamic acid, [4-(2,5-dioxo-4-oxazolidinyl)butyl]-, benzyl ester, L- (8CI), Carbamic acid, [4-(2,5-dioxo-4-oxazolidinyl)butyl]-, phenylmethyl ester, (S)- (9CI), N-ε-Benzyloxycarbonyl-L-lysine-N-carboxyanhydride, N6-Benzyloxycarbonyl-L-lysine N-carboxyanhydride, N6-Carbobenzoxy L-lysine N-carboxyanhydride, N6-Carbobenzyloxy-L-lysine N-carboxyanhydride, NCA-N-ε-benzyloxycarbonyl-L-lysine, Nε-Benzyloxycarbonyl-L-lysine N-carboxyanhydride, Nε-Benzyloxycarbonyl-L-lysine carboxyanhydride, Nε-Carbobenzoxylysine N-carboxy anhydride, Nε-Cbz-L-Lysine Nα-carboxyanhydride, Z-L-Lysine N-carboxyanhydride, ε-Benzyloxycarbonyl-L-lysine N-carboxyanhydride |
| InChI formel | InChI=1S/C15H18N2O5/c18-13-12(17-15(20)22-13)8-4-5-9-16-14(19)21-10-11-6-2-1-3-7-11/h1-3,6-7,12H,4-5,8-10H2,(H,16,19)(H,17,20)/t12-/m0/s1 |
| SMIL | O=C(NCCCC[C@@H]1NC(=O)OC1=O)OCc2ccccc2 |
| IUPAC navn | benzyl N-[4-[(4S)-2,5-dioxo-1,3-oxazolidin-4-yl]butyl]carbamate |
| Molekylvægt (g/mol) | 306.31 |
| Formel vægt | 306.1216 |