Få mere at vide
N4-Isobutylquinoline-3,4-diamine, Mikromol™
Artikelnummer. 30462188
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462188 4 x 25 mg
30051071 25 mg
30049378 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462188 Leverandør Mikromol Leverandørnr. MM1158.0600254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N4-Isobutylquinoline-3,4-diamine, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Isobutylamino-3-aminoquinoline
CAS 99010-09-0
Analyt- eller komponentnavne 4-Isobutylamino-3-aminoquinoline
Forsendelsestilstand Room Temperature
Molekylær formel C13 H17 N3
Certificeringer/compliance ISO 17025
Synonym 3,4-Quinolinediamine, N4-(2-methylpropyl)-, 3-Amino-4-(isobutylamino)quinoline, 4-Isobutylamino-3-aminoquinoline, N'-(2-Methylpropyl)quinoline-3,4-diamine, Imiquimod Related Compound E
InChI formel InChI=1S/C13H17N3/c1-9(2)7-16-13-10-5-3-4-6-12(10)15-8-11(13)14/h3-6,8-9H,7,14H2,1-2H3,(H,15,16)
Løsningstype Neat
SMIL CC(C)CNc1c(N)cnc2ccccc12
IUPAC navn 4-N-(2-methylpropyl)quinoline-3,4-diamine
Molekylvægt (g/mol) 215.29
Mængde 4 x 25 mg
Formel vægt 215.1422 g/mol