Få mere at vide
N4-Isobutylquinoline-3,4-diamine Hydrochloride, Mikromol™
Artikelnummer. 30052668
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052668 25 mg
30050140 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052668 Leverandør Mikromol Leverandørnr. MM1158.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N4-Isobutylquinoline-3,4-diamine Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N4-Isobutylquinoline-3,4-diamine Hydrochloride
CAS 935521-01-0
Analyt- eller komponentnavne N4-Isobutylquinoline-3,4-diamine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C13 H17 N3 . Cl H
Certificeringer/compliance ISO 17025
Synonym N4-Isobutylquinoline-3,4-diamine Hydrochloride, 3,4-Quinolinediamine, N4-(2-methylpropyl)-, hydrochloride (1:1), Imiquimod Related Compound E as Hydrochloride
InChI formel InChI=1S/C13H17N3.ClH/c1-9(2)7-16-13-10-5-3-4-6-12(10)15-8-11(13)14;/h3-6,8-9H,7,14H2,1-2H3,(H,15,16);1H
SMIL Cl.CC(C)CNc1c(N)cnc2ccccc12
IUPAC navn 4-N-(2-methylpropyl)quinoline-3,4-diamine;hydrochloride
Molekylvægt (g/mol) 251.76
Mængde 25 mg
Formel vægt 251.1189 g/mol