Få mere at vide
N3-(Benzyl-D7)-Adenine, TRC
Artikelnummer. 30347314
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30347314 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347314 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2244451MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N3-(Benzyl-D7)-Adenine
CAS 147028-87-3
Molekylær formel C12 D7 H4 N5
Mængde 1 mg
Synonym 6[2H7]benzylaminopurine, 2H-BA, 3-(Phenyl-d5-methyl-d2)-3H-purin-6-amine
InChI formel InChI=1S/C12H11N5/c13-11-10-12(15-7-14-10)17(8-16-11)6-9-4-2-1-3-5-9/h1-5,7-8H,6,13H2/i1D,2D,3D,4D,5D,6D2
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])n2cnc(N)c3ncnc23
IUPAC navn 3-[dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methyl]purin-6-amine
Molekylvægt (g/mol) 232.29
Formel vægt 232.1454