Få mere at vide
N2-(S)-1-Carboxy-3-phenylpropyl-L-lysine, TRC
Artikelnummer. 30004890
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004890 50 mg
17963745 100 mg
17993505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004890 Leverandør Toronto Research Chemicals Leverandørnr. TRCC18122550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 138247-43-5
Molekylær formel C16 H24 N2 O4
Molekylvægt (g/mol) 308.37
Synonym Benzenebutanoic acid, α-[[(1S)-5-amino-1-carboxypentyl]amino]-, (αS)-, Benzenebutanoic acid, α-[(5-amino-1-carboxypentyl)amino]-, [S-(R*,R*)]-, (αS)-α-[[(1S)-5-Amino-1-carboxypentyl]amino]benzenebutanoic acid, Lisinopril Dihydrate Imp. H (EP)
IUPAC navn (2S)-6-amino-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]hexanoic acid
SMIL NCCCC[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale N2-(S)-1-Carboxy-3-phenylpropyl-L-lysine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C16H24N2O4/c17-11-5-4-8-13(15(19)20)18-14(16(21)22)10-9-12-6-2-1-3-7-12/h1-3,6-7,13-14,18H,4-5,8-11,17H2,(H,19,20)(H,21,22)/t13-,14-/m0/s1
Mængde 50 mg
Formel vægt 308.1736
Procent renhed >95%