Få mere at vide
N2,N6-Bis(benzyloxycarbonyl)-L-lysine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N,N'-Bis(benzyloxycarbonyl)-L-lysine |
| CAS | 405-39-0 |
| Molekylær formel | C22 H26 N2 O6 |
| Mængde | 5 g |
| Synonym | L-Lysine, N2,N6-bis[(phenylmethoxy)carbonyl]-, Lysine, N2,N6-dicarboxy-, N2,N6-dibenzyl ester, L- (6CI,7CI,8CI), N2,N6-Bis[(phenylmethoxy)carbonyl]-L-lysine, 124: PN: WO2004099237 PAGE: 36 claimed protein, N,N'-Bis(benzyloxycarbonyl)-L-lysine, N,N'-Bis(benzyloxycarbonyl)lysine, N,N'-Di-Z-L-lysine, N,N'-Dicarbobenzoxy-L-lysine, N2,N6-Bis(benzyloxycarbonyl)-L-lysine, N2,N6-Dicarbobenzoxy-L-lysine, NSC 88474, Nα,Nε-Bis(benzyloxycarbonyl)lysine, Nα,Nε-Bis(carbobenzyloxy)-L-lysine, Nα,Nω-Bis(benzyloxycarbonyl)-L-lysine, Z-Lyz(Z)-OH, α,ε-Dicarbobenzoxy-L-lysine |
| InChI formel | InChI=1S/C22H26N2O6/c25-20(26)19(24-22(28)30-16-18-11-5-2-6-12-18)13-7-8-14-23-21(27)29-15-17-9-3-1-4-10-17/h1-6,9-12,19H,7-8,13-16H2,(H,23,27)(H,24,28)(H,25,26)/t19-/m0/s1 |
| SMIL | OC(=O)[C@H](CCCCNC(=O)OCc1ccccc1)NC(=O)OCc2ccccc2 |
| IUPAC navn | (2S)-2,6-bis(phenylmethoxycarbonylamino)hexanoic acid |
| Molekylvægt (g/mol) | 414.45 |
| Formel vægt | 414.1791 |