Få mere at vide
N2',N2' ' '-[Dithiobis(1-oxo-3,1-propanediyl)]bis[N2'-deacetyl- maytansine, TRC
Artikelnummer. 30011168
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30011168 25 mg -
30008389 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011168 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49418525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 905449-88-9
Molekylær formel C70H94Cl2N6O20S2
Molekylvægt (g/mol) 1474.56
Synonym DM1 Dimer
SMIL O=C(C[C@H](OC([C@H](C)N(C)C(CCSSCCC(N(C)[C@@H](C)C(O[C@H]([C@@]([C@H]1[C@@H]2C)(O1)C)CC(N(C)C3=CC(C/C(C)=C/C=C/[C@@H](OC)[C@]4(O)NC(O[C@H]2C4)=O)=CC(OC)=C3Cl)=O)=O)=O)=O)=O)[C@@]([C@H]5[C@@H]6C)(O5)C)N(C)C7=CC(C/C(C)=C/C=C/[C@@H](OC)[C@]8(O)NC(O[C@H]6C8)=O)=CC(OC)=C7Cl
Tekniske data
Kemisk navn eller materiale N2',N2' ' '-[Dithiobis(1-oxo-3,1-propanediyl)]bis[N2'-deacetyl-maytansine
Mængde 25 mg
InChI formel InChI=1S/C70H94Cl2N6O20S2/c1-37-19-17-21-51(91-15)69(87)35-49(93-65(85)73-69)39(3)61-67(7,97-61)53(33-57(81)77(11)45-29-43(27-37)31-47(89-13)59(45)71)95-63(83)41(5)75(9)55(79)23-25-99-100-26-24-56(80)76(10)42(6)64(84)96-54-34-58(82)78(12)46-30-44(32-48(90-14)60(46)72)28-38(2)20-18-22-52(92-16)70(88)36-50(94-66(86)74-70)40(4)62-68(54,8)98-62/h17-22,29-32,39-42,49-54,61-62,87-88H,23-28,33-36H2,1-16H3,(H,73,85)(H,74,86)/b21-17+,22-18+,37-19+,38-20+/t39-,40-,41+,42+,49+,50+,51-,52-,53+,54+,61-,62-,67+,68+,69+,70+/m1/s1