Få mere at vide
N2-Lauroyl-L-glutamine-d23, TRC
Artikelnummer. 30470248
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30470248 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470248 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17865210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N2-Lauroyl-L-glutamine-d23
CAS 1795786-88-7
Molekylær formel C17H9D23N2O4
Mængde 10 mg
Synonym N2-(1-Oxododecyl-d23)-L-glutamine
InChI formel InChI=1S/C17H32N2O4/c1-2-3-4-5-6-7-8-9-10-11-16(21)19-14(17(22)23)12-13-15(18)20/h14H,2-13H2,1H3,(H2,18,20)(H,19,21)(H,22,23)/t14-/m0/s1/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)N[C@@H](CCC(=O)N)C(=O)O
IUPAC navn (2S)-5-amino-5-oxo-2-(2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosadeuteriododecanoylamino)pentanoic acid
Molekylvægt (g/mol) 351.59
Formel vægt 351.3806