Få mere at vide
N1-Nornicergoline, TRC
Artikelnummer. 30134051
Skift visning
Mængde:
2.5 mg
5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30134051 2.5 mg 2.5mg
30134319 5 mg -
17913505 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134051 Leverandør Toronto Research Chemicals Leverandørnr. TRCN7566002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 35264-46-1
Molekylær formel C23 H24 Br N3 O3
Molekylvægt (g/mol) 470.36
Synonym Ergoline-8-methanol, 10-methoxy-6-methyl-, 8-(5-bromo-3-pyridinecarboxylate), (8β)-, Ergoline-8-methanol, 10-methoxy-6-methyl-, 5-bromo-3-pyridinecarboxylate (ester), (8β)- (9CI), 1-Demethylnicergoline, N1-Demethylnicergoline, N1-Nornicergoline, [(6aR,9R,10aS)-10a-Methoxy7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methyl 5-bromopyridine-3-carboxylate, 1-Desmethylnicergoline, Nicergoline Imp. B (EP)
IUPAC navn [(6aR,9R,10aS)-10a-methoxy-7-methyl-4,6,6a,8,9,10-hexahydroindolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
SMIL CO[C@]12C[C@@H](COC(=O)c3cncc(Br)c3)CN(C)[C@@H]1Cc4c[nH]c5cccc2c45
Tekniske data
Kemisk navn eller materiale 1-Demethylnicergoline
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C23H24BrN3O3/c1-27-12-14(13-30-22(28)16-6-17(24)11-25-9-16)8-23(29-2)18-4-3-5-19-21(18)15(10-26-19)7-20(23)27/h3-6,9-11,14,20,26H,7-8,12-13H2,1-2H3/t14-,20-,23+/m1/s1
Formel vægt 469.1001
Procent renhed >95%