missing translation for 'onlineSavingsMsg'
Få mere at vide

N1,N8-Diacetylspermidine, TRC

Artikelnummer. 30138446
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138446 1 g
30004025 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30138446 Leverandør Toronto Research Chemicals Leverandørnr. TRCD3670001G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82414-35-5
Molekylær formel C11H23N3O2
Molekylvægt (g/mol) 229.32
Synonym N-[3-[[4-(Acetylamino)butyl]amino]propyl]acetamide, N1,8-Diacetylspermidine
SMIL O=C(C)NCCCCNCCCNC(C)=O

Specifications

Kemisk navn eller materiale N1,N8-Diacetylspermidine
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C11H23N3O2/c1-10(15)13-8-4-3-6-12-7-5-9-14-11(2)16/h12H,3-9H2,1-2H3,(H,13,15)(H,14,16)
Formel vægt 229.179
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.