missing translation for 'onlineSavingsMsg'
Learn More

N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine, 98%, Thermo Scientific Chemicals

Catalog Number p-7033223
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
6342-21-8
C10H16N2
164.252
MFCD01631934
NFSAPTWLWWYADB-UHFFFAOYSA-N
2-amino-1-phenylethyl dimethylamine, n1,n1-dimethyl-1-phenyl-1,2-ethanediamine, n1,n1-dimethyl-1-phenylethane-1,2-diamine, n*1*,n*1*-dimethyl-1-phenyl-ethane-1,2-diamine, n∼1∼,n∼1∼-dimethyl-1-phenylethane-1,2-diamine, 1,2-ethanediamine, n1,n1-dimethyl-1-phenyl, acmc-20apq4, daa-0-0, n,n-dimethyl-1-phenyl-ethane-1,2-diamine
241652
N,N-dimethyl-1-phenylethane-1,2-diamine
CN(C)C(CN)C1=CC=CC=C1
This item is not returnable. View return policy
6342-21-8
C10H16N2
164.252
MFCD01631934
NFSAPTWLWWYADB-UHFFFAOYSA-N
2-amino-1-phenylethyl dimethylamine, n1,n1-dimethyl-1-phenyl-1,2-ethanediamine, n1,n1-dimethyl-1-phenylethane-1,2-diamine, n*1*,n*1*-dimethyl-1-phenyl-ethane-1,2-diamine, n∼1∼,n∼1∼-dimethyl-1-phenylethane-1,2-diamine, 1,2-ethanediamine, n1,n1-dimethyl-1-phenyl, acmc-20apq4, daa-0-0, n,n-dimethyl-1-phenyl-ethane-1,2-diamine
241652
N,N-dimethyl-1-phenylethane-1,2-diamine
CN(C)C(CN)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2735
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'