Få mere at vide
N1-Methylguanosine, TRC
Artikelnummer. 30008014
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008014 10 mg
30007024 2 mg
30006644 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008014 Leverandør Toronto Research Chemicals Leverandørnr. TRCB42659310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2140-65-0
Molekylær formel C11H15N5O5
Molekylvægt (g/mol) 297.27
Synonym 1-Methylguanosine
IUPAC navn 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1-methylpurin-6-one
SMIL CN1C(=O)C2=C(N=C1N)N(C=N2)[C@H]3[C@@H]([C@@H]([C@H](O3)CO)O)O
Tekniske data
Kemisk navn eller materiale N1-Methylguanosine
Mængde 10 mg
InChI formel InChI=1S/C11H15N5O5/c1-15-9(20)5-8(14-11(15)12)16(3-13-5)10-7(19)6(18)4(2-17)21-10/h3-4,6-7,10,17-19H,2H2,1H3,(H2,12,14)/t4-,6-,7-,10-/m1/s1
Formel vægt 297.1073