Få mere at vide
N1-(2-Aminophenyl)-N2,N2-dimethylglycinamide, TRC
Artikelnummer. 30337831
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30337831 250 mg
30333509 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30337831 Leverandør Toronto Research Chemicals Leverandørnr. TRCB406243250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N1-(2-Aminophenyl)-N2,N2-dimethylglycinamide
CAS 35266-48-9
Molekylær formel C10H15N3O
Mængde 250 mg
InChI formel InChI=1S/C10H15N3O/c1-13(2)7-10(14)12-9-6-4-3-5-8(9)11/h3-6H,7,11H2,1-2H3,(H,12,14)
SMIL CN(C)CC(=O)Nc1ccccc1N
IUPAC navn N-(2-aminophenyl)-2-(dimethylamino)acetamide
Molekylvægt (g/mol) 193.25
Formel vægt 193.1215