missing translation for 'onlineSavingsMsg'
Få mere at vide

N1-[2-[3,4-Bis(phenylmethoxy)phenyl]ethyl]-N6-(2-phenylethyl)-hexanediamide, TRC

Artikelnummer. 30337097
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30337097 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337097 Supplier Toronto Research Chemicals Supplier No. TRCB52142050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N1-[2-[3,4-Bis(phenylmethoxy)phenyl]ethyl]-N6-(2-phenylethyl)-hexanediamide
CAS 1357466-55-7
Molekylær formel C36H40N2O4
Mængde 50 mg
InChI formel InChI=1S/C36H40N2O4/c39-35(37-24-22-29-12-4-1-5-13-29)18-10-11-19-36(40)38-25-23-30-20-21-33(41-27-31-14-6-2-7-15-31)34(26-30)42-28-32-16-8-3-9-17-32/h1-9,12-17,20-21,26H,10-11,18-19,22-25,27-28H2,(H,37,39)(H,38,40)
SMIL O=C(CCCCC(=O)NCCc1ccc(OCc2ccccc2)c(OCc3ccccc3)c1)NCCc4ccccc4
IUPAC navn N'-[2-[3,4-bis(phenylmethoxy)phenyl]ethyl]-N-(2-phenylethyl)hexanediamide
Molekylvægt (g/mol) 564.71
Formel vægt 564.2988
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.