Få mere at vide
n-Undecyl-1,1,2,2-d4 Alcohol, CDN
Artikelnummer. 30318903
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318903 0.25 g
30321219 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318903 Leverandør CDN Leverandørnr. CDND67500.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Undecyl-1,1,2,2-d4 Alcohol
CAS 1219803-47-0
Molekylær formel C11 D4 H20 O
Mængde 0.25 g
Synonym 1-Undecan-1,1,2,2-d4-ol, Undecanol-1,1,2,2-d4, n-Undecyl-1,1,2,2-d4 alcohol, 1-Undecanol-1,1,2,2-d4, 1-Undecanol-1,1,2,2-D4, 1-Hydroxyundecane-1,1,2,2-d4
InChI formel InChI=1 S/C11H24O/c1-2-3-4-5-6-7-8-9-10-11-12/h12H,2-11H2,1H3/i10D2,11D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])CCCCCCCCC
IUPAC navn 1,1,2,2-tetradeuterioundecan-1-ol
Molekylvægt (g/mol) 176.332
Formel vægt 176.208 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity