Få mere at vide
N-Trityl Olmesartan Ethyl Ester, TRC
Artikelnummer. 30455536
Skift visning
Mængde:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455536 10 mg
30451092 100 mg
30455337 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30455536 Leverandør Toronto Research Chemicals Leverandørnr. TRCT88942510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 172875-59-1
Molekylær formel C45 H44 N6 O3
Molekylvægt (g/mol) 716.87
Synonym 1H-Imidazole-5-carboxylic acid, 4-(1-hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-, ethyl ester, 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid Ethyl Ester
IUPAC navn ethyl 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate
SMIL CCCc1nc(c(C(=O)OCC)n1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7)C(C)(C)O
Tekniske data
Kemisk navn eller materiale 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid Ethyl Ester
Mængde 10 mg
InChI formel InChI=1S/C45H44N6O3/c1-5-18-39-46-41(44(3,4)53)40(43(52)54-6-2)50(39)31-32-27-29-33(30-28-32)37-25-16-17-26-38(37)42-47-49-51(48-42)45(34-19-10-7-11-20-34,35-21-12-8-13-22-35)36-23-14-9-15-24-36/h7-17,19-30,53H,5-6,18,31H2,1-4H3
Formel vægt 716.3475