Få mere at vide
N-Trityl Losartan-d4, TRC
Artikelnummer. 30450355
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30450355 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450355 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8888221MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Trityl Losartan-d4
CAS 1309283-24-6
Molekylær formel C41H33D4ClN6O
Mængde 1 mg
Synonym 2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl-d4]-4-yl]methyl]-1H-imidazole-5-methanol, Trityllosartan-d4
InChI formel InChI=1S/C41H37ClN6O/c1-2-3-23-38-43-39(42)37(29-49)47(38)28-30-24-26-31(27-25-30)35-21-13-14-22-36(35)40-44-46-48(45-40)41(32-15-7-4-8-16-32,33-17-9-5-10-18-33)34-19-11-6-12-20-34/h4-22,24-27,49H,2-3,23,28-29H2,1H3/i24D,25D,26D,27D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1Cn2c(CCCC)nc(Cl)c2CO)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7
IUPAC navn [2-butyl-5-chloro-3-[[2,3,5,6-tetradeuterio-4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
Molekylvægt (g/mol) 669.25
Formel vægt 668.2968