Få mere at vide
N-Trityl Candesartan Cilexetil, TRC
Artikelnummer. 30456113
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456113 1 g
30453089 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30456113 Leverandør Toronto Research Chemicals Leverandørnr. TRCT8872251G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 886999-34-4
Molekylær formel C52 H48 N6 O6
Molekylvægt (g/mol) 852.97
Synonym 1H-Benzimidazole-7-carboxylic acid, 2-ethoxy-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, N2-Tritylcandesartan Cilexetil, 1-(cyclohexoxycarbonyloxy)ethyl 2-ethoxy-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
IUPAC navn 1-cyclohexyloxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
SMIL CCOc1nc2cccc(C(=O)OC(C)OC(=O)OC3CCCCC3)c2n1Cc4ccc(cc4)c5ccccc5c6nnn(n6)C(c7ccccc7)(c8ccccc8)c9ccccc9
Tekniske data
Kemisk navn eller materiale N2-Tritylcandesartan Cilexetil
Mængde 1 g
InChI formel InChI=1S/C52H48N6O6/c1-3-61-50-53-46-30-18-29-45(49(59)62-36(2)63-51(60)64-42-25-14-7-15-26-42)47(46)57(50)35-37-31-33-38(34-32-37)43-27-16-17-28-44(43)48-54-56-58(55-48)52(39-19-8-4-9-20-39,40-21-10-5-11-22-40)41-23-12-6-13-24-41/h4-6,8-13,16-24,27-34,36,42H,3,7,14-15,25-26,35H2,1-2H3
Formel vægt 852.3635