Få mere at vide
N-Trifluoroacetodesmethyl Citalopram-d3, TRC
Artikelnummer. 30454178
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30454178 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454178 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7863471MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Trifluoroacetodesmethyl Citalopram-d3
CAS 1189981-17-6
Molekylær formel C21H15D3F4N2O2
Mængde 1 mg
InChI formel InChI=1S/C21H18F4N2O2/c1-27(19(28)21(23,24)25)10-2-9-20(16-4-6-17(22)7-5-16)18-8-3-14(12-26)11-15(18)13-29-20/h3-8,11H,2,9-10,13H2,1H3/i1D3
SMIL [2H]C([2H])([2H])N(CCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3)C(=O)C(F)(F)F
IUPAC navn N-[3-[5-cyano-1-(4-fluorophenyl)-3H-2-benzofuran-1-yl]propyl]-2,2,2-trifluoro-N-(trideuteriomethyl)acetamide
Molekylvægt (g/mol) 409.39
Formel vægt 409.1493