Learn More
GPR63 Antibody, PerCP, R&D Systems™
Product Code. 18612058 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. Unit Size
18612058 0.1mL
1 options
Product Code. 18612058 Supplier Novus Biologicals Supplier No. NBP306018PCP
 Restricted Item
Forventet forsendelse: 10-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
GPR63 Polyclonal specifically detects GPR63 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Immunohistochemistry (Paraffin)/Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Gene Alias brain expressed G-protein-coupled receptor PSP24 beta, G protein-coupled receptor 63, probable G-protein coupled receptor 63, PSP24-2, PSP24-beta, PSP24BPSP24(beta)
Primary or Secondary Primary
Product Type Antibody
Product Line Novus